Stampa…

4-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected 98%

Fornitore: Apollo Scientific
Attenzione

Synonyms: 4-(Benzyloxy)-3-bromo-1-(tert-butoxycarbonyl)-1H-indole, tert-Butyl 4-(benzyloxy)-3-bromo-1H-indole-1-carboxylate

OR1718-1G OR1718-250MG
APOSOR1718-1GEA 270 EUR
APOSOR1718-1G APOSOR1718-250MG
4-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected 98%
4-(Benzyloxy)-3-bromo-1H-indole, N-BOC protected
Formula: C₂₀H₂₀BrNO₃
Peso molecolare: 402,282 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05864768
CAS Number: 914349-26-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR