Stampa…

(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid

Fornitore: Apollo Scientific

OR16060-1G OR16060-5G
APOSOR16060-1GEA 299 EUR
APOSOR16060-1G APOSOR16060-5G
(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid
(1R,4S)-Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid
Formula: C₁₀H₁₂O₄
Peso molecolare: 196,2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00267156
CAS Number: 34487-58-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR