Stampa…

5-(Hydroxymethyl)-3-methylisoxazole

Fornitore: Apollo Scientific

Synonyms: (3-Methyl-1,2-oxazol-5-yl)methanol, (3-Methylisoxazol-5-yl)methanol

OR15967-1G OR15967-5G
APOSOR15967-1GEA 147 EUR
APOSOR15967-1G APOSOR15967-5G
5-(Hydroxymethyl)-3-methylisoxazole
5-(Hydroxymethyl)-3-methylisoxazole
Formula: C₅H₇NO₂
Peso molecolare: 113,115 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09859210
CAS Number: 14716-89-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR