Cyclohexylmethanesulphonyl chloride
Fornitore: Apollo Scientific
|
Pericolo
|
Formula:
C₇H₁₃ClO₂S Punto di ebollizione: 92…95 °C (0,5 mm Hg) Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD03452748 CAS Number: 4352-30-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...