5-Bromo-2-hydrazino-3-nitropyridine
Fornitore: Apollo Scientific
|
Attenzione
|
OR11958-1G
OR11958-250MG
APOSOR11958-1GEA
48.1
EUR
APOSOR11958-1G
APOSOR11958-250MG
5-Bromo-2-hydrazino-3-nitropyridine
5-Bromo-2-hydrazino-3-nitropyridine
Formula:
C₅H₅BrN₄O₂ Peso molecolare: 233,023 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09801053 CAS Number: 15862-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...