6-Bromo-2-chloro-3-methylbenzeneboronic acid 97%
Fornitore: Apollo Scientific
Formula:
C₇H₇BBrClO₂ Punto di fusione: 170…175 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09475910 CAS Number: 957120-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...