Stampa…

5-Oxo-1-(2-thien-2-ylethyl)pyrrolidine-3-carboxylic acid

Fornitore: Apollo Scientific

OR111394-1G OR111394-5G
APOSOR111394-1GEA 227 EUR
APOSOR111394-1G APOSOR111394-5G
5-Oxo-1-(2-thien-2-ylethyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(2-thien-2-ylethyl)pyrrolidine-3-carboxylic acid
Formula: C₁₁H₁₃NO₃S
Peso molecolare: 239,291 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01934740
CAS Number: 261363-53-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR