Stampa…

Ethyl 4-iodo-5-methylisoxazole-3-carboxylate

Fornitore: Apollo Scientific
Attenzione

OR111318-1G OR111318-250MG
APOSOR111318-1GEA 226 EUR
APOSOR111318-1G APOSOR111318-250MG
Ethyl 4-iodo-5-methylisoxazole-3-carboxylate
Ethyl 4-iodo-5-methylisoxazole-3-carboxylate
Formula: C₇H₈INO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28992249
CAS Number: 1356600-24-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR