N-(4-Chloro-3-nitrophenyl)-N'-(4-chlorophenyl)urea
Fornitore: Apollo Scientific
Formula:
C₁₃H₉Cl₂N₃O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00126339 CAS Number: 304453-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...