N-(1,1,3,3,6-Pentamethyl-7-nitro-2,3-dihydro-1H-inden-5-yl)acetamide
Fornitore: Apollo Scientific
Formula:
C₁₆H₂₂N₂O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD12406814 CAS Number: 500282-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...