Stampa…

2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine

Fornitore: Apollo Scientific
Attenzione

Synonyms: 1,6-Dimethyl-1H-imidazo[4,5-b]pyridin-2-amine, DMIP

OR0518T-100MG OR0518T-10MG OR0518T-5MG
APOSOR0518T-100MGEA 2120 EUR
APOSOR0518T-100MG APOSOR0518T-10MG APOSOR0518T-5MG
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine

A mutagenic cogener of PHIP.

Formula: C₈H₁₀N₄
Peso molecolare: 162,192 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00269879
CAS Number: 132898-04-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR