Stampa…

N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride 97%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Piperazino-acetic acid-benzylamide dihydrochloride

OR0245-250MG OR0245-500MG
APOSOR0245-250MGEA 14.7 EUR
APOSOR0245-250MG APOSOR0245-500MG
N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride 97%
N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride
Formula: C₁₃H₁₉N₃O·2ClH
Peso molecolare: 306,231 g/mol
Punto di fusione: 218…221 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04115628
CAS Number: 827614-58-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR