Stampa…

Ethyl hept-6-enoate 97%

Fornitore: Apollo Scientific

OR01811-1G OR01811-5G
APOSOR01811-1GEA 125 EUR
APOSOR01811-1G APOSOR01811-5G
Ethyl hept-6-enoate 97%
Ethyl hept-6-enoate
Formula: C₉H₁₆O₂
Peso molecolare: 156,222 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00671845
CAS Number: 25118-23-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR