Stampa…

3-(1H-Indol-2-yl)aniline

Fornitore: Apollo Scientific

Synonyms: 2-(3-Aminophenyl)-1H-indole

OR01694-5G OR01694-1G
APOSOR01694-5GEA 823 EUR
APOSOR01694-5G APOSOR01694-1G
3-(1H-Indol-2-yl)aniline
3-(1H-Indol-2-yl)aniline
Formula: C₁₄H₁₂N₂
Peso molecolare: 208,258 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00454976
CAS Number: 6318-72-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR