7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
YN1
OR015000-1G
OR015000-250MG
APOSOR015000-1GEA
78.6
EUR
APOSOR015000-1G
APOSOR015000-250MG
7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
7,8-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Formula:
C₁₅H₁₀O₅ Peso molecolare: 270,237 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00885567 CAS Number: 75187-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...