Amino-PEG1-t-Butyl ester
Fornitore: Apollo Scientific
Synonyms:
tert-Butyl 3-(2-aminoethoxy)propanoate
BIPG1071-250MG
BIPG1071-1G
BIPG1071-500MG
APOSBIPG1071-250MGEA
118
EUR
APOSBIPG1071-250MG
APOSBIPG1071-1G
APOSBIPG1071-500MG
Amino-PEG1-t-Butyl ester
Amino-PEG1-t-Butyl ester
Formula:
C₉H₁₉NO₃ Peso molecolare: 189,252 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20727480 CAS Number: 1260092-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...