Stampa…

2'-Deoxyisoguanosine

Fornitore: Apollo Scientific

BIMN2012-100MG BIMN2012-250MG BIMN2012-500MG
APOSBIMN2012-100MGEA 348 EUR
APOSBIMN2012-100MG APOSBIMN2012-250MG APOSBIMN2012-500MG
2'-Deoxyisoguanosine
2'-Deoxyisoguanosine
Formula: C₁₀H₁₃N₅O₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD15145285
CAS Number: 106449-56-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR