Stampa…

5-Bromo-6-chloro-3-indolyl α-D-glucopyranoside

Fornitore: Apollo Scientific
Attenzione

Synonyms: Magenta-α-glucoside

BIMB3104-100MG BIMB3104-1G BIMB3104-250MG
APOSBIMB3104-100MGEA 65.5 EUR
APOSBIMB3104-100MG APOSBIMB3104-1G APOSBIMB3104-250MG
5-Bromo-6-chloro-3-indolyl α-D-glucopyranoside
5-Bromo-6-chloro-3-indolyl α-D-glucopyranoside
Formula: C₁₄H₁₅BrClNO₆
Peso molecolare: 408,629 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06796940
CAS Number: 878495-64-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR