5-Bromo-6-chloroindolyl-1,3-diacetate
Fornitore: Apollo Scientific
|
Attenzione
|
BIMB1194-1G
BIMB1194-250MG
APOSBIMB1194-1GEA
79.9
EUR
APOSBIMB1194-1G
APOSBIMB1194-250MG
5-Bromo-6-chloroindolyl-1,3-diacetate
5-Bromo-6-chloroindolyl-1,3-diacetate
Formula:
C₁₂H₉BrClNO₃ Peso molecolare: 330.562 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06797509 CAS Number: 108847-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...