Stampa…

N-Hydroxysulphosuccinimide

Fornitore: Apollo Scientific
Attenzione

Synonyms: Sulpho NHS

BIM108-500MG BIM108-1G BIM108-5G
APOSBIM108-500MGEA 87.3 EUR
APOSBIM108-500MG APOSBIM108-1G APOSBIM108-5G
N-Hydroxysulphosuccinimide
N-Hydroxysulphosuccinimide

Forms amine reactive peptides


  • Protein


Ref: Staros,J.V,Wright,R.W.and Swingle,D.M.(1986) Anal.Biochem.156, 220-222

Formula: C₄H₅NO₆S
Peso molecolare: 195,151 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00217065
CAS Number: 82436-78-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR