Stampa…

(S)-Fmoc-α-methyl-phenylglycine

Fornitore: Apollo Scientific
Attenzione

Synonyms: (S)-Fmoc-Mpg-OH, (2S)-2-{[(9H-Fluoren-9-ylmethoxy)carbonyl]amino}-2-phenylpropanoic acid

BICR127-1G BICR127-500MG
APOSBICR127-1GEA 578 EUR
APOSBICR127-1G APOSBICR127-500MG
(S)-Fmoc-α-methyl-phenylglycine
(S)-Fmoc-α-methyl-phenylglycine
Formula: C₂₄H₂₁NO₄
Peso molecolare: 387,428 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD11052918
CAS Number: 881921-11-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR