Stampa…

(S)-β3-phenylalanine

Fornitore: Apollo Scientific

BICR106-1G BICR106-5G BICR106-10G
APOSBICR106-1GEA 43.5 EUR
APOSBICR106-1G APOSBICR106-5G APOSBICR106-10G
(S)-β3-phenylalanine
(S)-β3-phenylalanine
Formula: C₉H₁₁NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01076238
CAS Number: 40856-44-8

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR