Stampa…

4-Methylphenyl 1-thio-β-D-glucopyranoside ≥99%

Fornitore: Apollo Scientific

Synonyms: Tolyl 1-thio-β-D-glucopyranoside, Thiocresyl β-D-glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(p-tolylthio)tetrahydro-2H-pyran-3,4,5-triol

BICL4145-1G BICL4145-5G BICL4145-2G
APOSBICL4145-1GEA 198 EUR
APOSBICL4145-1G APOSBICL4145-5G APOSBICL4145-2G
4-Methylphenyl 1-thio-β-D-glucopyranoside ≥99%
4-Methylphenyl 1-thio-β-D-glucopyranoside
Formula: C₁₃H₁₈O₅S
Peso molecolare: 286,344 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11045365
CAS Number: 1152-39-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR