Punicalin
Fornitore: Apollo Scientific
Formula:
C₃₄H₂₂O₂₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09838018 CAS Number: 65995-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...