Pinacyanol iodide 97%, Sigma-Aldrich®
Fornitore: SIGMA ALDRICH MICROSCOPY
|
Pericolo
|
M-Clarity™ quality level = MQ100
Formula:
C₂₅H₂₅IN₂ Punto di fusione: 295 °C (dec., lit.) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00011975 CAS Number: 605-91-4 EINECS: 210-099-4 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | Dark green to black powder, crystals or Crystalline powder |
Infrared spectrum | Conforms to structure. |
Uv-visible spectrum | In ethanol |
Carbon | 60.3%-64.7% |
Nitrogen | 5.5%- 6.1% |
Thin-layer | Consistent with 97% purity |
Solubility | 10 mg/ml(1%), ethanol; dark blue solution |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...