Betahistine dihydrochloride ≥98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
- Bioactive
Formula:
C₈H₁₂N₂·2HCl Punto di fusione: 150…154 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00012813 CAS Number: 5579-84-0 |
Risultati del test di specificazione
Appearance | White to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 150 to 154 °C |
Loss on drying | ≤1 % (1 g, 105 °C) |
Solubility | (5 g in 50 ml water) Clear solution |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...