Kartogenin
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
2-((4-Phenylphenyl)carbamoyl)benzoic acid, 2-([1,1'-Biphenyl]-4-ylcarbamoyl)benzoic acid, 2-(N-(4-Phenylphenyl)carbamoyl)benzoic acid, N-Biphenyl-4-yl-phthalamic acid
464660010
464662500
ACRO464660010EA
402
EUR
ACRO464660010
ACRO464662500
Kartogenin
Kartogenin
- Bioactive
Formula:
C₂₀H₁₅NO₃ Peso molecolare: 317,344 g/mol Punto di fusione: >280 °C (dec.) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00560510 CAS Number: 4727-31-5 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to light yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...