Olanzapine
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
- Bioactive
Formula:
C₁₇H₂₀N₄S Punto di fusione: 194 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00866702 CAS Number: 132539-06-1 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 192 to 196 °C |
Water | ≤1 % |
HPLC | ≥97.5 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...