Stampa…

Valnemulin hydrochloride

Fornitore: Thermo Fisher Scientific
Attenzione

462830010 462832500
ACRO462830010EA 280 EUR
ACRO462830010 ACRO462832500
Valnemulin hydrochloride
Valnemulin hydrochloride

  • Bioactive

Formula: C₃₁H₅₂N₂O₅S·HCl
Peso molecolare: 601.28 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD08458958
CAS Number: 133868-46-9
UN: 3077
ADR: 9 III

Visualizza i codici

Risultati del test di specificazione

Appearance White to yellow Powder
Infrared spectrum Conforms
Titration Argentometric 95.0 to 102.0 %
Water ≤4.0 %
HPLC ≥95.0 %
Specific optical rotation (Dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR