Stampa…
Lab installations for SCAT SafetyCaps
Fornitore: S.C.A.T.
160145 106406 117096 106502 108278 107836 502021 106677 307508 502049 160206
SCTV160145EA 48.6 EUR
SCTV160145 554-4356 SCTV117096 SCTV106502 SCTV108278 SCTV107836 SCTV502021 SCTV106677 SCTV307508 SCTV502049 SCTV160206
Lab installations for SCAT SafetyCaps
Forniture per cromatografia Pezzi di ricambio per cromatografia
SCAT lab installations can be completely integrated into lab furniture and offer a gastight system for safe fluid management.
  • Useful accessories for mounting SymLine installations
  • Technical devices and components for the use of level controls
  • Spare parts for ensuring the functionality of the SymLine system
SCAT lab installations fit any SCAT SafetyCap or SafetyWasteCap with GL 45 thread size, to effectively prevent health hazards and environmental damage. Changeable exhaust filter and tube connectors can be fixed directly onto the desktop forMore Product Information