Stampa…
Laboratory freezers
Fornitore: Liebherr
LG5216 LGN2323 LGN2723 LGX823
471-0655EA 0 EUR
471-0655 471-0657 471-0658 471-1095
Laboratory freezers
Congelatori Congelatori per usi generici/di laboratorio
Laboratory freezers with white epoxy stainless steel body.
  • Electronic regulation with LCD screen
  • Audible door alarm and temperature alarm with contact for remote alarm
  • Alarm memory; RS485 connection for control and traceability on PC
  • Reversible door
Side access port, 30 mm in diameter, for monitoring performance and qualification.
Housing for 8 mm diameter PT100 probe (probe not supplied) for centralised temperature management.
Temperature adjustable: from −18 to −28 °C.
Temperature: −20 °C ±5 °C Cofrac certified.