Stampa…
Raccordi per agitatori a globo, BOLA
Fornitore: Bohlender
C398-12 C399-12 C398-08 C393-12 C398-04
441-0076EA 158 EUR
441-0076 441-0084 BOHLC398-08 BOHLC393-12 BOHLC398-04
Raccordi per agitatori a globo, BOLA
Dispositivi agitatori Accessori di agitazione
These POM couplings are ideal for balancing misalignment of axes between stirrer and stirrer shaft. Suitable for glass, metal or Bola stirrer shafts. Both models are suitable for rotational speeds of up to 1200 rpm and can be used for both right and left-handed rotation.
  • Excellent chemical resistance, temperature resistance from –20 to +110 °C
  • Assembled by means of screw joints with clamp rings
  • Pivot (length 90 mm) can be shortened by the user
  • Maximum misalignment of 10 mm
Powerful transmission of up to 300 Ncm.
Powerful transmission of up to 300 Ncm.