Stampa…
Rain jackets, classic F440, Iona™
Fornitore: Portwest
F440NARXXL F440NARXXXL F440NARL F440NARS F440NARXL F440NARM
PRTWF440NARXXLEA 28.6 EUR
PRTWF440NARXXL PRTWF440NARXXXL PRTWF440NARL PRTWF440NARS PRTWF440NARXL PRTWF440NARM
Rain jackets, classic F440, Iona™
Abbigliamento da lavoro
Lightweight and practical the classic Iona rain jacket provides waterproof protection and increased visibility. Features include pack-away hood, adjustable drawcord hem, large storage pockets and vented back yoke and eyelets added breathability and comfort.
  • Made up of 100% Polyester, PVC Coated
  • Lightweight, waterproof fabric with taped seams prevents water penetration
  • Taped seams to provide additional protection
  • Reflective tape for increased visibility
  • Two pockets for secure storage
  • Front zip opening for easy access
Inner elasticated cuff. Retail bagMore Product Information
Visualizza i codici
Specifiche
Modello Classic F440
Applicazioni per abiti Industrial
Environmentally Preferable
Risk area Rain (waterproof)
Genere Unisex
Sleeve or leg length Long
Tipo di chiusura Con chiusura a zip
Tessuto 100% Polyester, PVC coated
Peso 210 g/m²