Stampa…
High visibility breathable trousers, S487
Fornitore: Portwest
S487YERM S487YERXXL S487YERS S487YERL S487YERXL
PRTWS487YERMEA 33.4 EUR
PRTWS487YERM PRTWS487YERXXL PRTWS487YERS PRTWS487YERL PRTWS487YERXL
High visibility breathable trousers, S487
Abbigliamento da lavoro
These breathable trousers have been designed using a waterproof and highly breathable fabric with taped seams to prevent water penetration. The lower leg zips complete with plackets allow for quick and easy fitting.
  • Shell fabric: 300D breathable 190 g
  • Reflective tape for increased visibility
  • Zipped ankles for easy fitting over work boots
  • 50+ UPF rated fabric to block 98% of UV rays
Extremely water resistant fabric finish, water beads away from fabric surface. Fully elasticated waistband for ultimate wearer comfort.
Certificazioni: Certified to EN ISO 20471 after 50x washes. Standards: RIS-3279-TOM ISSUE 1 (ORANGE ONLY), EN 343:2019 Class 4:3 X WP 10,000mm, MVP 10,000g/m²/24hrs, ANSI/ISEA 107: 2015 SUPPLEMENTAL CLASS E, EN ISO 20471 Class 1
Attenzione: Wash with like colours. Do not dry clean.