Stampa…
Chemical resistant gloves, Nitrosafe Plus, A812
Fornitore: Portwest
A812GNRL A812GNRM A812GNRS A812GNRXL A812GNRXS A812GNRXXL
PRTWA812GNRLEA 46.2 EUR
PRTWA812GNRL PRTWA812GNRM PRTWA812GNRS PRTWA812GNRXL PRTWA812GNRXS PRTWA812GNRXXL
Chemical resistant gloves, Nitrosafe Plus, A812
Guanti
This nitrile gauntlet is tested and certified to offer protection against many common chemicals found in industry today.
  • Anti-slip pattern offers high dexterity in wet and dry conditions
  • Ultra-sensitive and free of soluble proteins help minimise the risk of allergic reaction
  • Textured pattern for enhanced grip
  • Flock lined for added comfort
Ideal for manufacturing, paint applications, electronics and glass handling where silicone is problematic. Suitable for a broad range of industries, including industrial cleaning, laboratory work, light assembly, aerospace and the chemical industry.
Certificazioni: EN ISO 374-5 micro organisms; EN 388:2016 (3101X); EN ISO 374-1:2016 Type A (AJKLMNOPST); Cat. III
Food contact approved