Stampa…
Cerio (III) solfato 99.9% (REO, rare earth oxide basis)
Fornitore: Thermo Scientific
Attenzione
Synonyms: Dicerium trisulphate
378660500 378662500
ACRO378660500EA 98.2 EUR
ACRO378660500 ACRO378662500
Cerio (III) solfato 99.9% (REO, rare earth oxide basis)
Cerio (III) solfato
Formula: Ce₂O₁₂S₃
Peso molecolare: 568,42 g/mol
Densità: 2,89 g/cm³ (20 °C)
CAS Number: 13454-94-9
EINECS: 236-644-6
Visualizza i codici
Appearance White to off-white Powder or crystals
Titration Complexometric 46.0 to 50.0 % (Ce)
Total conc. of imp.as REO ≤0.1 %