Tetrapropylammonium perruthenate ≥98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
TPAP, Tetra-n-propylammonium perruthenate(VII)
Purity: 98%
CAS No.: 114615-82-6
Molecular Formula: C12H28NO4Ru
Formula Weight: 351.43
Storage Temperature: 2°C to 8°C
Melting Point: 160° dec.
UN No.: 1479
MDL No.: MFCD00074914
Formula:
C₁₂H₂₈NO₄Ru Punto di fusione: 160 °C (dec.) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00074914 CAS Number: 114615-82-6 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...