Stampa…
Calcio fosfato tribasico, EMPROVE® EXPERT BP, E341, Ph. Eur. excipient_suitable, SAFC®
Fornitore: MERCK PRODUCTION CHEMICALS
Synonyms: Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic
1.02143.1000 1.02143.9026 1.02143.9025
1.02143.1000EA 1320 EUR
1.02143.1000 1.02143.9026 1.02143.9025
Calcio fosfato tribasico, EMPROVE® EXPERT BP, E341, Ph. Eur. excipient_suitable, SAFC®
Calcio fosfato tribasico
Formula: Ca₃(PO₄)₂
Peso molecolare: 310.18 g/mol
Densità: 3.14 g/cm³ (20 °C)
MDL Number: MFCD00015984
CAS Number: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741