Stampa…
Lithium dihydrogen orthophosphate ≥97%
Fornitore: Thermo Scientific
Attenzione
Synonyms: Lithium dihydrogen phosphate, Lithium phosphate monobasic, Lithium dihydrogenorthophosphate, Lithium dihydrogenphosphate
A16987.0B A16987.30
A16987.0BEA 190 EUR
A16987.0B A16987.30
Lithium dihydrogen orthophosphate ≥97%
Lithium dihydrogen orthophosphate
Purity: 97% CAS No.: 13453-80-0 Molecular Formula: LiH2PO4 Formula Weight: 103.93 Odour: Odourless Melting Point: 100° MDL No.: MFCD00016188
Formula: H₂LiO₄P
Peso molecolare: 103,93 g/mol
Punto di fusione: >100 °C
Densità: 2,461
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016188
CAS Number: 13453-80-0
EINECS: 236-633-6