Stampa…
Industrial gloves, AlphaTec® 87-137
Fornitore: Ansell
87-137/8.5 87-137/6.5
ANSE87-137/8.5EA 35 EUR
ANSE87-137/8.5 ANSE87-137/6.5
Industrial gloves, AlphaTec® 87-137
Guanti
Unlined natural rubber gloves with excellent dexterity and tactility.
  • Beaded cuff for easy donning /doffing
  • Super thin and highly flexible
  • High resistance to water based chemicals
  • Diamond pattern on palms and fingers for excellent wet and dry grip
Recommended for: Fruit and vegetable processing, food processing and meat packing applications, maintenance inspection, laboratory maintenance and clean up, chemical handling, chemical processing and preparation.
Certificazioni: EN 388:2016 (X010A), EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:2016 VIRUS, EN 421; Cat. III
Food contact approved
Confezione: 12 pairs per bag.