Stampa…

1-Butylcyclohexanol ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 1-n-Butylcyclohexanol

B20579.06 B20579.14
B20579.06EA 57.5 EUR
B20579.06 B20579.14
1-Butylcyclohexanol ≥98%
1-Butylcyclohexanol

Purity: 98% CAS No.: 5445-30-7 Molecular Formula: C10H20O Formula Weight: 156.27 MDL No.: MFCD00021404

Formula: C₁₀H₂₀O
Peso molecolare: 156,27 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00021404
CAS Number: 5445-30-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR