Stampa…
(Methylsulphonyl)benzene 99%
Fornitore: Thermo Scientific
Attenzione
Synonyms: Methyl phenyl sulphone
353040050 353040250
ACRO353040050EA 45.8 EUR
ACRO353040050 ACRO353040250
(Methylsulphonyl)benzene 99%
(Methylsulphonyl)benzene
Formula: C₇H₈O₂S
Peso molecolare: 156,21 g/mol
Punto di fusione: 85…88 °C
MDL Number: MFCD00014741
CAS Number: 3112-85-4
EINECS: 221-476-8
Visualizza i codici
Appearance White Crystals
Infrared spectrum Conforms
Melting point 85 to 88 °C
GC ≥98.5 %