Zirconium(IV) butoxide ∼80% in 1-butanol
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Zirconium(IV) n-butoxide, Zirconium(IV) Tetrabutoxide
Z0016-500G
Z0016-25G
TCIAZ0016-500GEA
133
EUR
TCIAZ0016-500G
TCIAZ0016-25G
Zirconium(IV) butoxide ∼80% in 1-butanol
Zirconium(IV) butoxide
Formula:
C₁₆H₃₆O₄Zr Peso molecolare: 383,68 g/mol Flash Pt: 38°C |
MDL Number:
MFCD00048765 CAS Number: 1071-76-7 EINECS: 213-995-3 UN: 1120 ADR: 3,III |
Risultati del test di specificazione
Appearance | White to Yellow to Green clear liquid |
Concentration (Chelometric Method) | 80.0 to 86.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...