Stampa…

(-)-Synephrine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (R)-(-)-4-[1-Hydroxy-2-(methylamino)ethyl]phenol, (R)-(-)-4-Hydroxy-alpha-(methylaminomethyl)benzyl alcohol, (R)-(-)-1-(4-Hydroxyphenyl)-2-(methylamino)ethanol

Y0012-1G Y0012-5G
TCIAY0012-1GEA 126 EUR
TCIAY0012-1G TCIAY0012-5G
(-)-Synephrine ≥98.0% (by HPLC, titration analysis)
(-)-Synephrine
Formula: C₉H₁₃NO₂
Peso molecolare: 167,21 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 614-35-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c171540a58cf6c7ef390a521fd2e717af6cd85e1&vid=c171540a58cf6c7ef390a521fd2e717af6cd85e1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -58.0 to -63.0 deg (C=2, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR