Ittrio ossido ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Yttrium oxide, Yttrium (III) oxide
Y0007-25G
Y0007-100G
TCIAY0007-25GEA
45
EUR
TCIAY0007-25G
TCIAY0007-100G
Ittrio ossido ≥97.0% (by titrimetric analysis)
Ittrio ossido
Formula:
Y₂O₃ Peso molecolare: 225,81 g/mol |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 Merck Index: 14,10107 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...