Yohimbine hydrochloride ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂₁H₂₆N₂O₃ |
MDL Number:
MFCD00012674 CAS Number: 65-19-0 EINECS: 200-600-4 UN: 1544 ADR: 6.1,II Merck Index: 14,10102 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +100 to +105 deg (C=1, H2O) |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...