Stampa…

o-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (1,2-Phenylenebis(methylene))diphosphonic acid

X0075-200MG X0075-1G
TCIAX0075-200MGEA 34 EUR
TCIAX0075-200MG TCIAX0075-1G
o-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)
o-Xylylenediphosphonic acid
Formula: C₈H₁₂O₆P₂
Peso molecolare: 266,13 g/mol
MDL Number: MFCD22200404
CAS Number: 42104-58-5
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b7bc3950f8a746a24bddbc6ffae9814d705eed&vid=c1b7bc3950f8a746a24bddbc6ffae9814d705eed&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR