Stampa…

p-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (1,4-Phenylenebis(methylene))diphosphonic acid

X0074-1G X0074-5G
TCIAX0074-1GEA 98 EUR
TCIAX0074-1G TCIAX0074-5G
p-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)
p-Xylylenediphosphonic acid
Formula: C₈H₁₂O₆P₂
Peso molecolare: 266,13 g/mol
Punto di fusione: 280°C
MDL Number: MFCD05663703
CAS Number: 4546-06-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e1cffe0491316728a576b155a384857b733372b&vid=0e1cffe0491316728a576b155a384857b733372b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR