Stampa…

Xylometazoline hydrochloride ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-(4-(tert-Butyl)-2,6-dimethylbenzyl)-4,5-dihydro-1H-imidazole hydrochloride, 2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline Hydrochloride

X0063-1G X0063-5G
TCIAX0063-1GEA 29 EUR
TCIAX0063-1G TCIAX0063-5G
Xylometazoline hydrochloride ≥98.0% (by HPLC, total nitrogen)
Xylometazoline hydrochloride
Formula: C₁₆H₂₄N₂
Peso molecolare: 280,84 g/mol
MDL Number: MFCD00238707
CAS Number: 1218-35-5
EINECS: 214-936-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,10086

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR