Xylazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₂H₁₆N₂S |
MDL Number:
MFCD00058196 CAS Number: 23076-35-9 EINECS: 245-417-0 UN: 2811 ADR: 6.1,III Merck Index: 14,10080 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...